Assay Details
Target Gene Details
Entrez Gene ID: | 107568 |
Gene Name: | WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
8030445B08Rik, AIP5, SDRP1, Tiul1 |
Location: |
Chr.4:19608296-19709004 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wwp1 | NM_001276292.1 | 24 | 4567 | NP_001263221.1 |
| NM_177327.5 | 25 | 4729 | NP_796301.2 | |
| NR_074089.1 | 24 | 4404 | ||
| NR_074090.1 | 25 | 4626 | ||
| AK033138.1 | 17 | 3543 | BAC28168.1 | |
| AK042570.1 | 1 | 1240 | ||
| AK086189.1 | 1 | 611 | ||
| AK140411.1 | 1 | 1705 | ||
| BC051405.1 | 4 | 2047 | AAH51405.1 | |
| BC150812.1 | 24 | 4564 |
More Information
Set Membership: |
|

Genomic Map