Assay Details
Target Gene Details
Entrez Gene ID: | 22608 |
Gene Name: | Y box protein 1 |
Gene Aliases: |
1700102N10Rik, C79409, EF1A, MSY1, Nsep1, YB-1, dbpB, mYB-1a |
Location: |
Chr.4:119277327-119294659 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybx1 | NM_011732.2 | 8 | 1846 | NP_035862.2 |
| AK044081.1 | 3 | 979 | BAC31766.1 | |
| BG066506.2 | 1 | 346 |

Genomic Map