Assay Details
Target Gene Details
Entrez Gene ID: | 231123 |
Gene Name: | HAUS augmin-like complex, subunit 3 |
Gene Aliases: |
D4S43h, D5H4S43, D5H4S43E |
Location: |
Chr.5:34153896-34169424 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Haus3 | NM_146159.1 | 2 | 212 | NP_666271.1 |
| AK049414.1 | 1 | 1182 | BAC33742.1 | |
| BC023882.1 | 2 | 244 | AAH23882.1 | |
| BC025497.1 | 2 | 230 | AAH25497.1 | |
| BC027393.1 | 2 | 212 | AAH27393.1 |
Target Gene Details
Entrez Gene ID: | 272158 |
Gene Name: | DNA polymerase N |
Gene Aliases: |
POL4P |
Location: |
Chr.5:34007179-34169526 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Poln | NM_001289803.1 | NP_001276732.1 | ||
| NM_001289804.1 | NP_001276733.1 | |||
| NM_181857.4 | NP_862905.1 | |||
| AK133063.1 | BAE21491.1 | |||
| AY135562.3 | AAN39837.2 | |||
| BC141101.1 | ||||
| BC145378.1 | ||||
| CJ083086.1 |
More Information
Set Membership: |
|

Genomic Map