Assay Details
Target Gene Details
Entrez Gene ID: | 68033 |
Gene Name: | cytochrome c oxidase assembly protein 19 |
Gene Aliases: |
1810074D02Rik, 2810437L13Rik |
Location: |
Chr.5:139337822-139345166 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox19 | NM_197980.1 | 3 | 671 | NP_932097.1 |
| AK002486.1 | 3 | 687 | ||
| AK003697.1 | 3 | 715 | ||
| AK007984.1 | 3 | 680 | ||
| AK013260.1 | 3 | 687 | ||
| AK013289.1 | 3 | 715 | ||
| AK148526.1 | 3 | 644 | ||
| BC031792.1 | 3 | 671 | AAH31792.1 |
More Information
Set Membership: |
|

Genomic Map