Assay Details
Target Gene Details
Entrez Gene ID: | 442834 |
Gene Name: | RIKEN cDNA D830031N03 gene |
Gene Aliases: |
Kiaa0754, mKIAA0754 |
Location: |
Chr.4:123403601-123411911 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| D830031N03Rik | NM_001167918.1 | 1 | 2392 | NP_001161390.1 |
| AK173019.1 | 1 | 662 | BAD32297.1 |
Target Gene Details
Entrez Gene ID: | 11426 |
Gene Name: | microtubule-actin crosslinking factor 1 |
Gene Aliases: |
ABP620, Acf7, Aclp7, MACF, R74989, mKIAA0465 |
Location: |
Chr.4:123349633-123687315 on Build GRCm38 |
Assay Gene Location: | Within Intron 57 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Macf1 | NM_001199136.1 | NP_001186065.1 | ||
| NM_001199137.1 | NP_001186066.1 | |||
| AF150755.1 | AAD32244.1 | |||
| AK149066.1 | ||||
| AK162442.1 | BAE36920.1 | |||
| DQ067088.1 | AAY78553.1 |
More Information
Set Membership: |
|

Genomic Map