Assay Details
Target Gene Details
Entrez Gene ID: | 230099 |
Gene Name: | carbonic anhydrase 9 |
Gene Aliases: |
CAIX, Ca9, MN/CA9 |
Location: |
Chr.4:43507026-43513810 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car9 | NM_139305.2 | 11 | 1383 | NP_647466.2 |
| AB086322.1 | 9 | 1087 | BAC00816.1 | |
| AJ245857.1 | 11 | 1385 | CAC80975.1 | |
| AK136579.1 | 11 | 1384 | BAE23060.1 | |
| AK137719.1 | 11 | 1385 | BAE23474.1 | |
| AK166403.1 | 8 | 812 | BAE38755.1 | |
| BC120544.1 | 11 | 1376 | ||
| BC120546.1 | 11 | 1376 | ||
| EF122497.1 | 9 | 1091 |

Genomic Map