Assay Details
Target Gene Details
Entrez Gene ID: | 12448 |
Gene Name: | cyclin E2 |
Gene Aliases: |
- |
Location: |
Chr.4:11189214-11204779 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccne2 | NM_001037134.2 | 12 | 1876 | NP_001032211.1 |
| NM_001282943.1 | 12 | 1789 | NP_001269872.1 | |
| NM_009830.3 | 12 | 1786 | NP_033960.1 | |
| AF091432.1 | 11 | 1681 | AAC80527.1 | |
| AK010929.1 | 12 | 1716 | ||
| AK011883.1 | 7 | 1348 | BAB27897.1 | |
| AK031116.1 | 12 | 1714 | ||
| AK037791.1 | 12 | 1716 | BAC29873.1 | |
| AK158720.1 | 12 | 2302 | BAE34625.1 | |
| AK167428.1 | 12 | 1716 | BAE39516.1 | |
| BC053727.1 | 11 | 1676 | AAH53727.1 |
Target Gene Details
Entrez Gene ID: | 72656 |
Gene Name: | integrator complex subunit 8 |
Gene Aliases: |
2810013E07Rik, AV063769, D130008D20Rik |
Location: |
Chr.4:11199158-11254266 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ints8 | NM_001159595.1 | 27 | 4062 | NP_001153067.1 |
| NM_178112.5 | NP_835213.3 | |||
| AK049513.1 | BAC33786.1 | |||
| AK140829.1 | 27 | 4057 | BAE24491.1 | |
| AK164951.1 | 7 | 1465 | BAE37977.1 |
More Information
Set Membership: |
|

Genomic Map