Assay Details
Target Gene Details
Entrez Gene ID: | 76574 |
Gene Name: | major facilitator superfamily domain containing 2A |
Gene Aliases: |
1700018O18Rik, Mfsd2, NLS1 |
Location: |
Chr.4:122946850-122961195 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd2a | NM_029662.2 | 14 | 1929 | NP_083938.2 |
| AK006096.1 | 14 | 1928 | BAB24407.1 | |
| BC046793.1 | 14 | 1911 | AAH46793.1 | |
| BC060526.1 | 14 | 1900 | AAH60526.1 |

Genomic Map