Assay Details
Target Gene Details
Entrez Gene ID: | 230233 |
Gene Name: | inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein |
Gene Aliases: |
3110040G09Rik, 6030413P05, C78473, Elp1, IKAP |
Location: |
Chr.4:56749680-56802331 on Build GRCm38 |
Assay Gene Location: | Within Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikbkap | NM_026079.3 | 37 | 5134 | NP_080355.2 |
| AK032563.1 | 4 | 1303 | ||
| AK076152.1 | 10 | 1836 | BAC36221.1 |
More Information
Set Membership: |
|

Genomic Map