Assay Details
Target Gene Details
Entrez Gene ID: | 668173 |
Gene Name: | peroxisomal biogenesis factor 10 |
Gene Aliases: |
AV128229, Gm142 |
Location: |
Chr.4:155067030-155072406 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex10 | NM_001042407.1 | 6 | 1059 | NP_001035866.1 |
| AK043682.1 | 4 | 2428 | ||
| BB265326.2 | 2 | 313 | ||
| BC147304.1 | 6 | 1044 | ||
| BC147305.1 | 6 | 1044 | ||
| BY385428.1 | ||||
| CB203398.1 | 5 | 633 |
More Information
Set Membership: |
|

Genomic Map