Assay Details
Target Gene Details
Entrez Gene ID: | 100764 |
Gene Name: | RBPJ interacting and tubulin associated 1 |
Gene Aliases: |
1110008J03Rik, AI853657, C86297, Rita |
Location: |
Chr.5:120609058-120612589 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rita1 | NM_029096.3 | 1 | 165 | NP_083372.1 |
| NM_133908.1 | 1 | 165 | NP_598669.1 | |
| AK003581.1 | 1 | 146 | BAB22871.1 | |
| BC021365.1 | 1 | 64 | AAH21365.1 | |
| CX239969.1 | 1 | 146 |

Genomic Map