Assay Details
Target Gene Details
Entrez Gene ID: | 230101 |
Gene Name: | glucosidase beta 2 |
Gene Aliases: |
F630034E04 |
Location: |
Chr.4:43566928-43578864 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gba2 | NM_172692.3 | 17 | 3082 | NP_766280.2 |
| AK089192.1 | 17 | 2768 | BAC40785.1 | |
| AK158174.1 | 15 | 3453 | ||
| AK173213.1 | 17 | 3068 | BAD32491.1 | |
| BC056935.1 | 17 | 2605 | AAH56935.1 |
More Information
Set Membership: |
|

Genomic Map