Assay Details
Target Gene Details
Entrez Gene ID: | 14590 |
Gene Name: | gamma-glutamyl hydrolase |
Gene Aliases: |
gamma-GH |
Location: |
Chr.4:20041987-20066111 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ggh | NM_010281.2 | 9 | 1136 | NP_034411.2 |
| AF051102.1 | 9 | 1104 | AAC70002.1 | |
| AI840300.1 | 1 | 205 | ||
| AK089996.1 | 9 | 1093 | ||
| BC009809.1 | 9 | 1027 | AAH09809.1 | |
| BC150898.1 | 9 | 1073 | ||
| CO044650.1 | 1 | 208 | ||
| EU694104.1 |
More Information
Set Membership: |
|

Genomic Map