Assay Details
Target Gene Details
Entrez Gene ID: | 14621 |
Gene Name: | gap junction protein, beta 4 |
Gene Aliases: |
Cnx30.3, Cx30.3, Gjb-4 |
Location: |
Chr.4:127331418-127354141 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjb4 | NM_008127.4 | 2 | 284 | NP_032153.1 |
| AK076408.1 | 2 | 285 | BAC36327.1 | |
| AK132468.1 | 2 | 281 | ||
| BC064060.1 | 1 | 222 | AAH64060.1 |
More Information
Set Membership: |
|

Genomic Map