Assay Details
Target Gene Details
Entrez Gene ID: | 230767 |
Gene Name: | IQ motif containing C |
Gene Aliases: |
BC026978 |
Location: |
Chr.4:129615617-129619213 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iqcc | NM_198026.2 | 5 | 1618 | NP_932143.2 |
| BC026978.1 | 3 | 1393 | AAH26978.1 |

Genomic Map