Assay Details
Target Gene Details
Entrez Gene ID: | 76804 |
Gene Name: | lysine (K)-specific demethylase 4C |
Gene Aliases: |
2410141F18Rik, AA517467, Gasc1, Jmjd2c |
Location: |
Chr.4:74242497-74405864 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm4c | NM_001172095.1 | 11 | 1621 | NP_001165566.1 |
| NM_144787.2 | 11 | 1685 | NP_659036.1 | |
| AK053104.1 | 11 | 1561 | BAC35267.1 | |
| AK081769.1 | 11 | 1628 | BAC38327.1 | |
| AK144103.1 | 11 | 1648 | BAE25700.1 | |
| AK144890.1 | 11 | 1659 | BAE26118.1 | |
| AK173023.1 | 11 | 1685 | BAD32301.1 | |
| BC020180.1 | 11 | 1627 | AAH20180.1 | |
| BC042424.1 | 11 | 1621 | AAH42424.1 |
More Information
Set Membership: |
|

Genomic Map