Assay Details
Target Gene Details
Entrez Gene ID: | 230801 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class V |
Gene Aliases: |
B330013B03, D430024F16Rik |
Location: |
Chr.4:133659914-133673997 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigv | NM_001145955.1 | 4 | 1189 | NP_001139427.1 |
| NM_001145956.1 | 3 | 913 | NP_001139428.1 | |
| NM_178698.5 | 4 | 2035 | NP_848813.3 | |
| AK046527.1 | 4 | 1996 | BAC32773.1 | |
| AK047322.1 | 4 | 1833 | BAC33023.1 | |
| AK052446.1 | 4 | 2009 | BAC34995.1 | |
| AK080590.1 | 3 | 914 | BAC37950.1 | |
| AK080858.1 | 4 | 1989 | BAC38047.1 | |
| AK148285.1 | 3 | 902 | BAE28458.1 | |
| BC055060.1 | 4 | 1985 | AAH55060.1 |
More Information
Set Membership: |
|

Genomic Map