Assay Details
Target Gene Details
Entrez Gene ID: | 13483 |
Gene Name: | dipeptidylpeptidase 6 |
Gene Aliases: |
B930011P16Rik, D5Buc3, D5Buc4, D5Buc5, DPP VI, Dpp-6, Gm1377, In(5)6H-p, Peplb, Rw |
Location: |
Chr.5:26817357-27727500 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpp6 | NM_010075.2 | 1 | 640 | NP_034205.1 |
| AF092507.1 | 1 | 648 | AAC97366.1 | |
| AK082186.1 | 1 | 564 | ||
| AK136075.1 | 1 | 795 |
More Information
Set Membership: |
|

Genomic Map