Assay Details
Target Gene Details
Entrez Gene ID: | 18198 |
Gene Name: | muscle, skeletal, receptor tyrosine kinase |
Gene Aliases: |
Mdk4, Mlk, Nsk1, Nsk2, Nsk3 |
Location: |
Chr.4:58285640-58377848 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Musk | NM_001037127.2 | 16 | 2363 | NP_001032204.2 |
| NM_001037128.1 | 16 | 2318 | NP_001032205.1 | |
| NM_001037129.1 | 15 | 2294 | NP_001032206.1 | |
| NM_001037130.1 | 14 | 2264 | NP_001032207.1 | |
| NM_010944.2 | 15 | 2288 | NP_035074.2 | |
| AB691772.1 | 14 | 2114 | ||
| AY955296.1 | 16 | 2213 | AAY40318.1 | |
| AY955297.1 | 16 | 2168 | AAY40319.1 | |
| AY955298.1 | 15 | 2144 | AAY40320.1 | |
| AY956403.1 | 15 | 2138 | AAY40321.1 | |
| AY956404.1 | 14 | 2114 | AAY40322.1 | |
| BC100342.1 | 15 | 2218 | AAI00343.1 | |
| U37708.1 | 14 | 2262 | AAA79203.1 | |
| U37709.1 | 15 | 2286 | AAA79204.1 | |
| X86444.1 | 14 | 2192 | CAA60165.1 |
More Information
Set Membership: |
|

Genomic Map