Assay Details
Target Gene Details
Entrez Gene ID: | 18780 |
Gene Name: | phospholipase A2, group IIA (platelets, synovial fluid) |
Gene Aliases: |
EF, Mom1, Pla2, sPLA2, sPla2-IIA |
Location: |
Chr.4:138831857-138835189 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g2a | NM_001082531.1 | 5 | 404 | NP_001076000.1 |
| NR_002926.3 | 5 | 424 | ||
| AK131857.1 | 5 | 425 | ||
| BC045156.1 | 5 | 404 | AAH45156.1 | |
| CB933893.1 | 2 | 116 | ||
| U28244.1 | 5 | 411 | AAB06315.1 | |
| U32359.1 | 4 | 379 | AAB60494.1 | |
| X74266.1 | 5 | 420 | CAA52325.1 |
Target Gene Details
Entrez Gene ID: | 18784 |
Gene Name: | phospholipase A2, group V |
Gene Aliases: |
PLA2, sPLA2 |
Location: |
Chr.4:138799247-138863469 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g5 | NM_001122954.1 | NP_001116426.1 | ||
| AK090021.1 | BAC41050.1 | |||
| BY012884.1 | ||||
| BY015217.1 |
More Information
Set Membership: |
|

Genomic Map