Assay Details
Target Gene Details
Entrez Gene ID: | 11781 |
Gene Name: | adaptor-related protein complex AP-4, mu 1 |
Gene Aliases: |
4930443L05Rik, Ap4m4 |
Location: |
Chr.5:138172021-138178687 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap4m1 | NM_021392.4 | 15 | 1449 | NP_067367.3 |
| AF242858.1 | 15 | 1317 | AAF63513.1 | |
| AK031638.1 | 15 | 1452 | BAC27490.1 | |
| BC011174.1 | 15 | 1334 | AAH11174.1 | |
| BE627979.1 | 1 | 287 |
More Information
Set Membership: |
|

Genomic Map