Assay Details
Target Gene Details
Entrez Gene ID: | 54170 |
Gene Name: | Ras-related GTP binding C |
Gene Aliases: |
AU041672, Gtr2, RAGC, TIB929, YGR163W |
Location: |
Chr.4:123917433-123936997 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rragc | NM_017475.2 | 7 | 2358 | NP_059503.2 |
| AB017616.1 | 7 | 2281 | BAA75671.1 | |
| AK003876.1 | 1 | 214 | ||
| AK150792.1 | 7 | 2313 | ||
| AK159355.1 | 7 | 2291 | BAE35013.1 | |
| AK159434.1 | 7 | 2313 | ||
| AK166658.1 | BAE38923.1 | |||
| AK177349.1 | 1 | 130 | ||
| AK196201.1 | 1 | 146 | ||
| AK196968.1 | 1 | 170 | ||
| AK203699.1 | 1 | 130 | ||
| AK204008.1 | 1 | 130 | ||
| AK213206.1 | 1 | 142 | ||
| BC005417.1 | 7 | 2306 | AAH05417.1 | |
| BC071245.1 | 7 | 2706 | AAH71245.1 | |
| BE628749.1 | 1 | 264 |
More Information
Set Membership: |
|

Genomic Map