Assay Details
Target Gene Details
Entrez Gene ID: | 12571 |
Gene Name: | cyclin-dependent kinase 6 |
Gene Aliases: |
5830411I20, AI504062, Crk2 |
Location: |
Chr.5:3343891-3531008 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk6 | NM_009873.2 | 7 | 2199 | NP_034003.1 |
| AF132483.1 | 7 | 2199 | AAD43504.1 | |
| AK154197.1 | 8 | 2472 | BAE32432.1 |
More Information
Set Membership: |
|

Genomic Map