Assay Details
Target Gene Details
Entrez Gene ID: | 209354 |
Gene Name: | eukaryotic translation initiation factor 2B, subunit 1 (alpha) |
Gene Aliases: |
26kDa, D5Ertd406e, EIF2B, EIF2BA |
Location: |
Chr.5:124570214-124579057 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2b1 | NM_145371.3 | 9 | 1555 | NP_663346.1 |
| AK133695.1 | 9 | 1556 | BAE21787.1 |
More Information
Set Membership: |
|

Genomic Map