Assay Details
Target Gene Details
Entrez Gene ID: | 100087 |
Gene Name: | KTI12 homolog, chromatin associated (S. cerevisiae) |
Gene Aliases: |
1110001A12Rik, AI326810, AU014758 |
Location: |
Chr.4:108847857-108849412 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kti12 | NM_029571.2 | 1 | 1037 | NP_083847.1 |
| AK003197.1 | 1 | 1038 | BAB22635.1 | |
| AK148185.1 | 1 | 1027 | BAE28403.1 | |
| AK150178.1 | 1 | 1025 | ||
| AK156755.1 | 1 | 1005 | BAE33838.1 | |
| BC021493.1 | 1 | 1120 | AAH21493.1 |
Target Gene Details
Entrez Gene ID: | 66073 |
Gene Name: | thioredoxin domain containing 12 (endoplasmic reticulum) |
Gene Aliases: |
0610040B21Rik, ERp16, ERp19 |
Location: |
Chr.4:108834678-108862120 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Txndc12 | NM_025334.3 | NP_079610.1 | ||
| AK002862.1 | BAB22413.1 | |||
| AK003481.1 | BAB22811.1 | |||
| AY548113.1 | AAS55653.1 | |||
| BC006857.1 | AAH06857.1 |
More Information
Set Membership: |
|

Genomic Map