Assay Details
Target Gene Details
Entrez Gene ID: | 12395 |
Gene Name: | runt-related transcription factor 1; translocated to, 1 (cyclin D-related) |
Gene Aliases: |
Cbfa2t1h, ETO, MTG8 |
Location: |
Chr.4:13743182-13895056 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Runx1t1 | NM_001111026.2 | NP_001104496.1 | ||
| NM_001111027.2 | 1 | 77 | NP_001104497.1 | |
| AK032132.1 | BAC27719.1 | |||
| AK142006.1 | BAE24910.1 | |||
| CB249744.1 | 1 | 77 |
More Information
Set Membership: |
|

Genomic Map