Assay Details
Target Gene Details
Entrez Gene ID: | 110326 |
Gene Name: | taste receptor, type 1, member 1 |
Gene Aliases: |
Gpr70, T1r1, TR1 |
Location: |
Chr.4:152027914-152038490 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas1r1 | NM_031867.2 | 6 | 2074 | NP_114073.1 |
| AF301161.1 | 6 | 1970 | AAK07091.1 | |
| AF301162.1 | 6 | 1970 | AAK07092.1 | |
| AF337040.1 | 6 | 2152 | AAK39437.1 | |
| AK153579.1 | 6 | 2024 | BAE32107.1 | |
| AY032622.1 | 6 | 1970 | AAK51603.1 |
More Information
Set Membership: |
|

Genomic Map