Assay Details
Target Gene Details
Entrez Gene ID: | 66729 |
Gene Name: | ankyrin repeat domain 61 |
Gene Aliases: |
4921520G13Rik |
Location: |
Chr.5:143890741-143895152 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ankrd61 | NM_025732.3 | 3 | 581 | NP_080008.1 |
AK014929.1 | 3 | 582 | BAB29624.1 | |
AK017053.1 | 3 | 522 | BAB30569.1 | |
BC052346.1 | 3 | 803 | AAH52346.1 |
