Assay Details
Target Gene Details
Entrez Gene ID: | 14228 |
Gene Name: | FK506 binding protein 4 |
Gene Aliases: |
59kDa, AL022792, AW208983, FKBP-4, FKBP-52, FKBP52, FKPB52, p59 |
Location: |
Chr.6:128430107-128438631 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp4 | NM_010219.3 | 10 | 2040 | NP_034349.1 |
| AK035395.1 | 2 | 730 | BAC29057.1 | |
| AK083912.1 | 10 | 2040 | BAC39057.1 | |
| AK089972.1 | 10 | 2049 | ||
| AK219397.1 | ||||
| X17069.2 | 10 | 1906 | CAC39452.1 | |
| X70887.1 | 10 | 1937 | CAA50231.1 |
More Information
Set Membership: |
|

Genomic Map