Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxp2 | NM_001286607.1 | 1 | 101 | NP_001273536.1 |
| NM_053242.4 | NP_444472.2 | |||
| NM_212435.1 | 1 | 101 | NP_997600.1 | |
| AK051668.1 | BAC34711.1 | |||
| AK080549.1 | ||||
| AK082361.1 | BAC38477.1 | |||
| AK164319.1 | BAE37735.1 | |||
| AY079003.1 | AAL85482.1 | |||
| BC058960.1 | 1 | 98 | AAH58960.1 | |
| BC062926.1 | 1 | 101 | AAH62926.1 |

Genomic Map