Assay Details
Target Gene Details
Entrez Gene ID: | 14823 |
Gene Name: | glutamate receptor, metabotropic 8 |
Gene Aliases: |
A230002O04, Gprc1h, mGluR8 |
Location: |
Chr.6:27275121-28135837 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm8 | NM_008174.2 | 11 | 3583 | NP_032200.2 |
| AK038395.1 | 11 | 3637 | ||
| AK134845.1 | 3 | 1550 | BAE22309.1 | |
| BC051384.1 | 11 | 3583 |
More Information
Set Membership: |
|

Genomic Map