Assay Details
Target Gene Details
Entrez Gene ID: | 100609 |
Gene Name: | NOL1/NOP2/Sun domain family, member 5 |
Gene Aliases: |
9830109N13Rik, AI326939, Nol1r, Wbscr20, Wbscr20a |
Location: |
Chr.5:135369930-135376805 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nsun5 | NM_145414.2 | 10 | 1619 | NP_663389.2 |
| AF412029.1 | AAM62311.1 | |||
| AK079400.1 | BAC37634.1 | |||
| AK149521.1 | 10 | 1601 | BAE28935.1 | |
| AK156651.1 | 10 | 1614 | BAE33793.1 | |
| BC051209.1 | AAH51209.1 |
Target Gene Details
Entrez Gene ID: | 107939 |
Gene Name: | nuclear pore membrane protein 121 |
Gene Aliases: |
2610027A18Rik, C80273, mKIAA0618 |
Location: |
Chr.5:135376140-135404733 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pom121 | NM_148932.2 | NP_683734.2 | ||
| AF516680.1 | AAM64199.1 | |||
| AK154473.1 | BAE32610.1 | |||
| AK172982.1 | BAD32260.1 | |||
| CF545814.1 | ||||
| CJ306468.1 |
More Information
Set Membership: |
|

Genomic Map