Assay Details
Target Gene Details
Entrez Gene ID: | 66596 |
Gene Name: | general transcription factor III A |
Gene Aliases: |
2010015D03Rik, 2610111I01Rik, 5330403M05Rik |
Location: |
Chr.5:146948657-146955614 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf3a | NM_025652.3 | 1 | 268 | NP_079928.2 |
| AF391799.1 | 1 | 252 | AAL69686.1 | |
| AK008247.2 | 1 | 234 | BAC25209.1 | |
| AK011845.1 | 1 | 229 | ||
| AK012797.1 | 1 | 215 | BAB28476.2 | |
| AK017234.1 | 1 | 163 | ||
| AK143627.1 | 1 | 220 | ||
| AK183221.1 | 1 | 49 | ||
| BE656279.1 | 1 | 268 |
More Information
Set Membership: |
|

Genomic Map