Assay Details
Target Gene Details
Entrez Gene ID: | 320405 |
Gene Name: | Ca2+-dependent activator protein for secretion 2 |
Gene Aliases: |
A230044C21Rik, Caps-2, Caps2, Cpd2 |
Location: |
Chr.6:23262773-23839421 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadps2 | NM_001252105.1 | 29 | 4501 | NP_001239034.1 |
| NM_001252106.1 | 29 | 4315 | NP_001239035.1 | |
| NM_001252110.1 | 28 | 4486 | NP_001239039.1 | |
| NM_153163.4 | 30 | 4621 | NP_694803.3 | |
| AB098623.1 | 29 | 4500 | BAD05017.1 | |
| AB291947.1 | 30 | 4436 | ||
| AB291948.1 | 29 | 4200 | ||
| AF000969.2 | 30 | 4247 | AAB58720.2 | |
| AI877214.1 | ||||
| AK038568.1 | 30 | 4426 | BAC30047.1 | |
| AK078741.1 | 12 | 1653 | ||
| AY072800.1 | 31 | 4250 | AAL67937.1 | |
| BB521660.2 | 1 | 580 | ||
| BC047394.1 | 28 | 4114 | AAH47394.1 | |
| BC059274.1 | 27 | 3835 | AAH59274.1 | |
| BC080854.1 | 29 | 3934 | AAH80854.1 | |
| CB233516.1 | 2 | 641 | ||
| EH103433.1 | 1 | 392 |
More Information
Set Membership: |
|

Genomic Map