Assay Details
Target Gene Details
Entrez Gene ID: | 15932 |
Gene Name: | iduronidase, alpha-L- |
Gene Aliases: |
6030426D08 |
Location: |
Chr.5:108660331-108685446 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idua | NM_008325.4 | NP_032351.2 | ||
| NM_172997.1 | NP_766585.1 | |||
| AK031414.1 | BAC27393.1 | |||
| AK045330.1 | BAC32313.1 | |||
| AK155627.1 | BAE33353.1 | |||
| AK169998.1 | BAE41504.1 | |||
| BC057935.1 | AAH57935.1 | |||
| L34111.1 | AAC42044.1 |
Target Gene Details
Entrez Gene ID: | 231583 |
Gene Name: | solute carrier family 26 (sulfate transporter), member 1 |
Gene Aliases: |
Sat1 |
Location: |
Chr.5:108669970-108678097 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a1 | NM_174870.3 | 1 | 350 | NP_777359.3 |
| AK085280.1 | 1 | 58 | ||
| AK128888.1 | 1 | 70 | ||
| AK143528.1 | 1 | 78 | BAE25419.1 | |
| AY093420.1 | 1 | 270 | AAM18183.1 | |
| BC022130.1 | AAH22130.1 | |||
| BC032151.1 | 1 | 78 | AAH32151.1 |
More Information
Set Membership: |
|

Genomic Map