Assay Details
Target Gene Details
Entrez Gene ID: | 74340 |
Gene Name: | S-adenosylhomocysteine hydrolase-like 2 |
Gene Aliases: |
4631427C17Rik, AI227036, Adohcyase3, mKIAA0828 |
Location: |
Chr.6:29768379-29912310 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahcyl2 | NM_001171000.2 | 1 | 150 | NP_001164471.2 |
| NM_021414.6 | 1 | 150 | NP_067389.5 | |
| AB041551.1 | 1 | 95 | BAA95036.1 | |
| AK014539.1 | 1 | 101 | ||
| AK134919.1 | 1 | 66 | BAE22337.1 | |
| AK165725.1 | 1 | 150 | BAE38354.1 | |
| BC027436.1 | 1 | 65 | ||
| BC079660.1 | 1 | 60 | AAH79660.1 | |
| BY733512.1 | 1 | 150 |
More Information
Set Membership: |
|

Genomic Map