Assay Details
Target Gene Details
Entrez Gene ID: | 74340 |
Gene Name: | S-adenosylhomocysteine hydrolase-like 2 |
Gene Aliases: |
4631427C17Rik, AI227036, Adohcyase3, mKIAA0828 |
Location: |
Chr.6:29768379-29912310 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahcyl2 | NM_001171000.2 | 17 | 2470 | NP_001164471.2 |
| NM_001171001.1 | 17 | 2190 | NP_001164472.1 | |
| NM_021414.6 | 17 | 2473 | NP_067389.5 | |
| AK014539.1 | 18 | 2424 | ||
| AK053527.1 | 17 | 2154 | BAC35415.1 | |
| AK122382.1 | 15 | 2176 | BAC65664.1 | |
| BC079660.1 | 17 | 2383 | AAH79660.1 |
More Information
Set Membership: |
|

Genomic Map