Assay Details
Target Gene Details
Entrez Gene ID: | 12386 |
Gene Name: | catenin (cadherin associated protein), alpha 2 |
Gene Aliases: |
AI481747, Catna, Catna2, cdf, chp |
Location: |
Chr.6:76881637-77979694 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctnna2 | NM_001109764.1 | NP_001103234.1 | ||
| NM_009819.2 | NP_033949.2 | |||
| NM_145732.2 | NP_663785.2 | |||
| AK158916.1 | BAE34726.1 | |||
| BC079648.1 | AAH79648.1 | |||
| D25281.1 | BAA04969.1 | |||
| D25282.1 | BAA04970.1 |
Target Gene Details
Entrez Gene ID: | 74342 |
Gene Name: | leucine rich repeat transmembrane neuronal 1 |
Gene Aliases: |
4632401D06Rik, AW125451 |
Location: |
Chr.6:77242480-77259915 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrtm1 | NM_028880.3 | 2 | 2182 | NP_083156.2 |
| AK014545.1 | 2 | 2185 | ||
| AK053345.1 | 2 | 2172 | BAC35354.1 | |
| AK053364.1 | 2 | 1806 | BAC35363.1 | |
| AK080836.1 | 2 | 2213 | ||
| AK136857.1 | 1 | 1681 | BAE23148.1 | |
| BC027803.1 | 2 | 2064 | AAH27803.1 | |
| BQ176603.1 | 1 | 355 |
More Information
Set Membership: |
|

Genomic Map