Assay Details
Target Gene Details
Entrez Gene ID: | 12468 |
Gene Name: | chaperonin containing Tcp1, subunit 7 (eta) |
Gene Aliases: |
AA408524, AL022769, Ccth, Cctz |
Location: |
Chr.6:85451505-85468477 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cct7 | NM_007638.4 | 1 | 418 | NP_031664.3 |
| BQ951185.1 | 1 | 418 |
Target Gene Details
Entrez Gene ID: | 73327 |
Gene Name: | protease-associated domain containing 1 |
Gene Aliases: |
1700040I03Rik, Pap21 |
Location: |
Chr.6:85446785-85451970 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pradc1 | NR_028081.1 | |||
| BG088874.2 | ||||
| CJ075544.1 |
More Information
Set Membership: |
|

Genomic Map