Assay Details
Target Gene Details
Entrez Gene ID: | 64177 |
Gene Name: | transient receptor potential cation channel, subfamily V, member 6 |
Gene Aliases: |
CAT, CaT1, Cac, Ecac2, Otrpc3 |
Location: |
Chr.6:41620619-41636480 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trpv6 | NM_022413.4 | 15 | 2317 | NP_071858.3 |
| AB037373.1 | 15 | 2224 | BAA99538.1 | |
| AJ542487.1 | 15 | 2022 | CAD62684.1 | |
| AK077234.1 | 15 | 2247 | BAC36699.1 | |
| AK168760.1 | 15 | 2315 | ||
| BC016101.1 | 15 | 2255 | AAH16101.1 | |
| CF583712.1 |
More Information
Set Membership: |
|

Genomic Map