Assay Details
Target Gene Details
Entrez Gene ID: | 264134 |
Gene Name: | tetratricopeptide repeat domain 26 |
Gene Aliases: |
9330141E21Rik, 9430097H08Rik |
Location: |
Chr.6:38381474-38427647 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc26 | NM_153600.2 | 18 | 3718 | NP_705828.2 |
| AK086670.1 | 2 | 2187 | BAC39716.1 | |
| BC031513.1 | 7 | 2121 | AAH31513.1 | |
| BC066060.1 | 18 | 3718 | AAH66060.1 |

Genomic Map