Assay Details
Target Gene Details
Entrez Gene ID: | 17135 |
Gene Name: | v-maf musculoaponeurotic fibrosarcoma oncogene family, protein K (avian) |
Gene Aliases: |
AW061068, NF-E2, Nfe2u |
Location: |
Chr.5:139791536-139802652 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mafk | NM_010757.2 | 3 | 1541 | NP_034887.1 |
| AK143654.1 | 3 | 1547 | BAE25480.1 | |
| AK143717.1 | 3 | 1545 | BAE25513.1 | |
| AK153856.1 | 3 | 1511 | BAE32212.1 | |
| BC014295.1 | 3 | 1519 | AAH14295.1 | |
| D42124.1 | 3 | 1544 | BAA07704.1 |
More Information
Set Membership: |
|

Genomic Map