Assay Details
Target Gene Details
Entrez Gene ID: | 109978 |
Gene Name: | ADP-ribosyltransferase 4 |
Gene Aliases: |
4432404K01Rik, ARTC4, DO, DOK1 |
Location: |
Chr.6:136848451-136857600 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Art4 | NM_026639.2 | 3 | 1350 | NP_080915.1 |
| AK004744.1 | 3 | 1304 | BAB23525.1 | |
| AK019467.1 | 3 | 1351 | BAB31738.1 | |
| BC046306.1 | 3 | 1271 | AAH46306.1 |
More Information
Set Membership: |
|

Genomic Map