Assay Details
Target Gene Details
Entrez Gene ID: | 50540 |
Gene Name: | immunoglobulin (CD79A) binding protein 1b |
Gene Aliases: |
alpha4-b |
Location: |
Chr.6:138657089-138658444 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igbp1b | NM_015777.2 | 1 | 912 | NP_056592.2 |
| AJ010637.1 | 1 | 912 | CAB60142.1 | |
| BC125601.1 | 1 | 1012 | ||
| BC131968.1 | 1 | 1012 |
More Information
Set Membership: |
|

Genomic Map