Assay Details
Target Gene Details
Entrez Gene ID: | 66999 |
Gene Name: | mediator complex subunit 28 |
Gene Aliases: |
1500003D12Rik, AI451633, AU045690, Eg1, FKSG20, magicin |
Location: |
Chr.5:45520229-45529284 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med28 | NM_025895.4 | 4 | 3188 | NP_080171.1 |
| AK154249.1 | 4 | 3114 | BAE32464.1 |
More Information
Set Membership: |
|

Genomic Map