Assay Details
Target Gene Details
Entrez Gene ID: | 80751 |
Gene Name: | ring finger protein 34 |
Gene Aliases: |
AW061037, AW536122, BC004042, C88279, RIFF |
Location: |
Chr.5:122850162-122868945 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf34 | NM_030564.1 | 3 | 400 | NP_085041.1 |
| AF434815.1 | 2 | 295 | AAL30770.1 | |
| AK045696.1 | 3 | 396 | BAC32461.1 | |
| AK046274.1 | 3 | 382 | BAC32666.1 | |
| AK046381.1 | 3 | 396 | BAC32697.1 | |
| AK077221.1 | 3 | 397 | BAC36693.1 | |
| AK150976.1 | 3 | 394 | BAE30005.1 | |
| AK159753.1 | 3 | 382 | BAE35344.1 | |
| BC004042.1 | 3 | 400 | AAH04042.1 |
More Information
Set Membership: |
|

Genomic Map