Assay Details
Target Gene Details
Entrez Gene ID: | 67855 |
Gene Name: | aspartic peptidase, retroviral-like 1 |
Gene Aliases: |
2300003P22Rik, AA986851, SASP, SASPase, Taps |
Location: |
Chr.6:86628174-86629710 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asprv1 | NM_026414.2 | 1 | 1270 | NP_080690.2 |
| AK004007.1 | 1 | 717 | BAB23121.2 | |
| AK160494.1 | 1 | 1212 | ||
| BC057938.1 | 1 | 1283 | AAH57938.1 | |
| BC108357.1 | 1 | 1168 | AAI08358.1 | |
| BC119099.1 | 1 | 786 | ||
| BC119101.1 | 1 | 786 | ||
| DQ841260.1 | 1 | 1270 |
More Information
Set Membership: |
|

Genomic Map