Assay Details
Target Gene Details
Entrez Gene ID: | 94047 |
Gene Name: | cat eye syndrome chromosome region, candidate 6 |
Gene Aliases: |
AW048664 |
Location: |
Chr.6:120488939-120493807 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cecr6 | NM_033567.1 | 1 | 3246 | NP_291045.1 |
| AF277399.1 | 1 | 3246 | AAK17095.1 | |
| AK140695.1 | 2 | 2316 | BAE24447.1 |

Genomic Map