Assay Details
Target Gene Details
Entrez Gene ID: | 70147 |
Gene Name: | RIKEN cDNA 2210019I11 gene |
Gene Aliases: |
- |
Location: |
Chr.5:147228598-147271521 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2210019I11Rik | NR_038157.1 | |||
| AK008755.1 | ||||
| AK133519.1 |
Target Gene Details
Entrez Gene ID: | 18609 |
Gene Name: | pancreatic and duodenal homeobox 1 |
Gene Aliases: |
IDX-1, IPF-1, Ipf1, Mody4, STF-1, pdx-1 |
Location: |
Chr.5:147270131-147275149 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdx1 | NM_008814.3 | 1 | 177 | NP_032840.1 |
| BC103572.1 | 1 | 177 | AAI03573.1 | |
| BC103581.1 | 1 | 177 | AAI03582.1 | |
| BC103582.1 | 1 | 177 | AAI03583.1 | |
| BC105642.1 | 1 | 177 | ||
| X74342.1 | 1 | 349 | CAA52389.1 |
More Information
Set Membership: |
|

Genomic Map