Assay Details
Target Gene Details
Entrez Gene ID: | 68770 |
Gene Name: | putative homeodomain transcription factor 2 |
Gene Aliases: |
1110054G21Rik, 9530062N20Rik, AI447096, C77923 |
Location: |
Chr.5:20758664-20882124 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phtf2 | NM_172992.3 | 18 | 4032 | NP_766580.3 |
| AK004246.1 | ||||
| AK151762.1 | 1 | 784 | ||
| BC052841.1 | 18 | 3945 | AAH52841.1 |
More Information
Set Membership: |
|

Genomic Map